Assay Details
| Gene Symbol: LOC105601854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012130401.3 | XP_011985791.2 | 1 | 485 | 109 | |
| Gene Symbol: GLRX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969160.1 | XP_027824961.1 | 3 | 570 | 109 | |
| XM_027969161.1 | XP_027824962.1 | 3 | 575 | 109 | ||
More Information
Set Membership: |
|

LOC105601854
Genomic Map