Assay Details
| Gene Symbol: LOC101116059 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972313.1 | XP_027828114.1 | 19 - 20 | 2030 | 70 | |
| XM_027972314.1 | XP_027828115.1 | 19 - 20 | 1853 | 70 | ||
| XM_027972315.1 | XP_027828116.1 | 18 - 19 | 1874 | 70 | ||
| XM_027972316.1 | XP_027828117.1 | 19 - 20 | 2047 | 70 | ||
| XM_027972317.1 | XP_027828118.1 | 18 - 19 | 1865 | 70 | ||
| XM_027972318.1 | XP_027828119.1 | 19 - 20 | 1880 | 70 | ||
| XM_027972319.1 | XP_027828120.1 | 19 - 20 | 1880 | 70 | ||
More Information
Set Membership: |
|

LOC101116059
Genomic Map