Assay Details
| Gene Symbol: LOC101117653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964755.1 | XP_027820556.1 | 13 - 14 | 1130 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117653| Gene Symbol: LOC101117653 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964755.1 | XP_027820556.1 | 13 - 14 | 1130 | 92 | |
Set Membership: |
|