Assay Details
| Gene Symbol: LOC101122274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096076.2 | XP_014951562.2 | 1 - 2 | 3036 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101122274| Gene Symbol: LOC101122274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096076.2 | XP_014951562.2 | 1 - 2 | 3036 | 77 | |
Set Membership: |
|