Assay Details
| Gene Symbol: LOC101102276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002607.4 | XP_004002656.2 | 2 - 3 | 287 | 99 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101102276| Gene Symbol: LOC101102276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002607.4 | XP_004002656.2 | 2 - 3 | 287 | 99 | |