Assay Details
| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002950.4 | XP_004002999.1 | 10 - 11 | 1461 | 124 | |
| XM_027980259.1 | XP_027836060.1 | 10 - 11 | 1461 | 124 | ||
More Information
Set Membership: |
|

CFAP91
Genomic Map