Assay Details
| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002950.4 | XP_004002999.1 | 1 - 2 | 246 | 69 | |
| XM_027980259.1 | XP_027836060.1 | 1 - 2 | 246 | 69 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP91| Gene Symbol: CFAP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002950.4 | XP_004002999.1 | 1 - 2 | 246 | 69 | |
| XM_027980259.1 | XP_027836060.1 | 1 - 2 | 246 | 69 | ||