Assay Details
| Gene Symbol: SWAP70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016163.3 | XP_004016212.2 | 9 - 10 | 1445 | 70 | |
| XM_027979400.1 | XP_027835201.1 | 8 - 9 | 1249 | 70 | ||
More Information
Set Membership: |
|

SWAP70
Genomic Map