Assay Details
Gene Symbol: LOC114117777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977532.1 | XP_027833333.1 | 3 - 4 | 772 | 67 |
Gene Symbol: LOC105601924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977510.1 | XP_027833311.1 | 3 - 4 | 745 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
