Assay Details
| Gene Symbol: LOC105604626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961389.1 | XP_027817190.1 | 4 - 5 | 224 | 95 | |
| XM_027961390.1 | XP_027817191.1 | 4 - 5 | 420 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105604626| Gene Symbol: LOC105604626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961389.1 | XP_027817190.1 | 4 - 5 | 224 | 95 | |
| XM_027961390.1 | XP_027817191.1 | 4 - 5 | 420 | 95 | ||