Assay Details
| Gene Symbol: LOC105604626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961389.1 | XP_027817190.1 | 6 - 7 | 301 | 67 | |
| XM_027961390.1 | XP_027817191.1 | 6 - 7 | 497 | 67 | ||
More Information
Set Membership: |
|

LOC105604626
Genomic Map