Assay Details
| Gene Symbol: CNTFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012117996.3 | XP_011973386.1 | 2 - 3 | 4591 | 66 | |
| XM_015093061.2 | XP_014948547.1 | 3 - 4 | 358 | 66 | ||
| XM_015093062.2 | XP_014948548.1 | 3 - 4 | 287 | 66 | ||
| XM_027964277.1 | XP_027820078.1 | 4 - 5 | 392 | 66 | ||
| XM_027964278.1 | XP_027820079.1 | 3 - 4 | 243 | 66 | ||
| XM_027964279.1 | XP_027820080.1 | 3 - 4 | 246 | 66 | ||
| XM_027964280.1 | XP_027820081.1 | 4 - 5 | 346 | 66 | ||
| XM_027964282.1 | XP_027820083.1 | 2 - 3 | 160 | 66 | ||
| XM_027964283.1 | XP_027820084.1 | 3 - 4 | 284 | 66 | ||
More Information
Set Membership: |
|

CNTFR
Genomic Map