Assay Details
| Gene Symbol: LOC101102529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961780.1 | XP_027817581.1 | 1 - 2 | 140 | 89 | |
| XM_027961781.1 | XP_027817582.1 | 1 - 2 | 140 | 89 | ||
| XM_027961782.1 | XP_027817583.1 | 1 - 2 | 140 | 89 | ||
More Information
Set Membership: |
|

LOC101102529
Genomic Map