Assay Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960660.1 | XP_027816461.1 | 3 - 4 | 1605 | 71 | |
| XM_027960661.1 | XP_027816462.1 | 4 - 5 | 2667 | 71 | ||
More Information
Set Membership: |
|

SLC16A12
Genomic Map