Assay Details
| Gene Symbol: SLC16A12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960660.1 | XP_027816461.1 | 4 - 5 | 2188 | 79 | |
| XM_027960661.1 | XP_027816462.1 | 5 - 6 | 3250 | 79 | ||
More Information
Set Membership: |
|

SLC16A12
Genomic Map