Assay Details
| Gene Symbol: LOC780488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959680.1 | XP_027815481.1 | 20 - 21 | 5452 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC780488| Gene Symbol: LOC780488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959680.1 | XP_027815481.1 | 20 - 21 | 5452 | 112 | |
Set Membership: |
|