Assay Details
| Gene Symbol: LOC114116929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975351.1 | XP_027831152.1 | 1 - 2 | 305 | 93 | |
| XM_027975352.1 | XP_027831153.1 | 2 - 3 | 603 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027975351.1, XM_027975352.1 transcripts.
Set Membership: |
|

LOC114116929
Genomic Map