Assay Details
| Gene Symbol: LOC114116929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975351.1 | XP_027831152.1 | 3 - 4 | 606 | 67 | |
| XM_027975352.1 | XP_027831153.1 | 4 - 5 | 904 | 67 | ||
More Information
Set Membership: |
|

LOC114116929
Genomic Map