Assay Details
| Gene Symbol: LOC114116929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975351.1 | XP_027831152.1 | 5 - 6 | 815 | 95 | |
| XM_027975352.1 | XP_027831153.1 | 6 - 7 | 1113 | 95 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114116929| Gene Symbol: LOC114116929 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975351.1 | XP_027831152.1 | 5 - 6 | 815 | 95 | |
| XM_027975352.1 | XP_027831153.1 | 6 - 7 | 1113 | 95 | ||