Assay Details
| Gene Symbol: NDUFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002954.4 | XP_004003003.2 | 1 - 2 | 219 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NDUFB4| Gene Symbol: NDUFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002954.4 | XP_004003003.2 | 1 - 2 | 219 | 105 | |
Set Membership: |
|