Assay Details
| Gene Symbol: TMEM252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004336.3 | XP_004004385.1 | 1 - 2 | 573 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM252| Gene Symbol: TMEM252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004336.3 | XP_004004385.1 | 1 - 2 | 573 | 88 | |
Set Membership: |
|