Assay Details
| Gene Symbol: OSGEPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004507.4 | XP_004004556.2 | 6 - 7 | 1312 | 65 | |
| XM_027964713.1 | XP_027820514.1 | 6 - 7 | 1096 | 65 | ||
More Information
Set Membership: |
|

OSGEPL1
Genomic Map