Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015209.4 | XP_004015258.2 | 2 - 3 | 384 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

THAP8| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015209.4 | XP_004015258.2 | 2 - 3 | 384 | 64 | |
Set Membership: |
|