Assay Details
| Gene Symbol: RPS19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015294.3 | XP_004015343.1 | 3 - 4 | 643 | 109 | |
| XM_027978401.1 | XP_027834202.1 | 3 - 4 | 190 | 109 | ||
More Information
Set Membership: |
|

RPS19
Genomic Map