Assay Details
| Gene Symbol: CHMP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015466.4 | XP_004015515.1 | 2 - 3 | 339 | 66 | |
| XM_004015467.4 | XP_004015516.1 | 2 - 3 | 372 | 66 | ||
| XM_027978944.1 | XP_027834745.1 | 2 - 3 | 303 | 66 | ||
More Information
Set Membership: |
|

CHMP2A
Genomic Map