Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017027.3 | XP_004017076.2 | 16 - 17 | 2523 | 112 | |
| XM_012097013.3 | XP_011952403.1 | 15 - 16 | 2384 | 112 | ||
| XM_012097014.2 | XP_011952404.1 | 16 - 17 | 2562 | 112 | ||
| XM_012097016.3 | XP_011952406.1 | 16 - 17 | 2738 | 112 | ||
More Information
Set Membership: |
|

FYB1
Genomic Map