Assay Details
| Gene Symbol: LOC114118096 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978907.1 | XP_027834708.1 | 3 - 4 | 460 | 111 | |
| Gene Symbol: LOC101107463 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027978908.1 | XP_027834709.1 | 3 - 4 | 470 | 111 | |
More Information
Set Membership: |
|

LOC114118096
Genomic Map