Assay Details
| Gene Symbol: WIPF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012133307.2 | XP_011988697.2 | 4 - 5 | 2478 | 81 | |
| XM_012133312.3 | XP_011988702.2 | 4 - 5 | 634 | 81 | ||
| XM_027964833.1 | XP_027820634.1 | 4 - 5 | 526 | 81 | ||
| XM_027964834.1 | XP_027820635.1 | 4 - 5 | 565 | 81 | ||
More Information
Set Membership: |
|

WIPF1
Genomic Map