Assay Details
| Gene Symbol: C24H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104074.2 | XP_014959560.2 | 1 - 2 | 624 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C24H16orf72| Gene Symbol: C24H16orf72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015104074.2 | XP_014959560.2 | 1 - 2 | 624 | 65 | |
Set Membership: |
|