Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015099129.2 | XP_014954615.2 | 3 - 4 | 2051 | 88 | |
| XM_027976208.1 | XP_027832009.1 | 4 - 5 | 1618 | 88 | ||
| XM_027976209.1 | XP_027832010.1 | 5 - 6 | 1717 | 88 | ||
| XM_027976210.1 | XP_027832011.1 | 2 - 3 | 1237 | 88 | ||
More Information
Set Membership: |
|

TMCC2
Genomic Map