Assay Details
Gene Symbol: LOC101110432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187503.3 | XP_012042893.1 | 4 - 5 | 1040 | 106 |

More Information
Set Membership: |
![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101110432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012187503.3 | XP_012042893.1 | 4 - 5 | 1040 | 106 |
Set Membership: |
![]() ![]() |