Assay Details
| Gene Symbol: MRTFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962081.1 | XP_027817882.1 | 16 - 17 | 2920 | 83 | |
| XM_027962082.1 | XP_027817883.1 | 14 - 15 | 2685 | 83 | ||
| XM_027962083.1 | XP_027817884.1 | 14 - 15 | 2829 | 83 | ||
| XM_027962084.1 | XP_027817885.1 | 14 - 15 | 2660 | 83 | ||
| XM_027962085.1 | XP_027817886.1 | 12 - 13 | 3929 | 83 | ||
| XM_027962086.1 | XP_027817887.1 | 16 - 17 | 2773 | 83 | ||
More Information
Set Membership: |
|

MRTFB
Genomic Map