Assay Details
| Gene Symbol: ARHGAP39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973332.1 | XP_027829133.1 | 9 - 10 | 3455 | 71 | |
| XM_027973333.1 | XP_027829134.1 | 10 - 11 | 3123 | 71 | ||
| XM_027973334.1 | XP_027829135.1 | 7 - 8 | 3556 | 71 | ||
| XM_027973335.1 | XP_027829136.1 | 9 - 10 | 3030 | 71 | ||
More Information
Set Membership: |
|

ARHGAP39
Genomic Map