Assay Details
| Gene Symbol: FBXO31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189651.2 | XP_012045041.2 | 5 - 6 | 768 | 59 | |
| XM_015100402.2 | XP_014955888.2 | 5 - 6 | 763 | 59 | ||
| XM_015100403.2 | XP_014955889.2 | 4 - 5 | 676 | 59 | ||
| XM_027977711.1 | XP_027833512.1 | 5 - 6 | 426 | 59 | ||
| XM_027977712.1 | XP_027833513.1 | 4 - 5 | 336 | 59 | ||
More Information
Set Membership: |
|

FBXO31
Genomic Map