Assay Details
| Gene Symbol: LOC101108006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002706.4 | XP_004002755.2 | 2 - 3 | 271 | 100 | |
| XM_012185108.3 | XP_012040498.1 | 2 - 3 | 271 | 100 | ||
More Information
Set Membership: |
|

LOC101108006
Genomic Map