Assay Details
Gene Symbol: CASR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956314.1 | XP_027812115.1 | 3 - 4 | 1493 | 84 | |
XM_027956319.1 | XP_027812120.1 | 3 - 4 | 1493 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
