Assay Details
| Gene Symbol: LOC114113045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964680.1 | XP_027820481.1 | 4 - 5 | 865 | 113 | |
| Gene Symbol: ERCC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964679.1 | XP_027820480.1 | 12 - 13 | 2007 | 113 | |
More Information
Set Membership: |
|

LOC114113045
Genomic Map