Assay Details
| Gene Symbol: LOC114113152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964842.1 | XP_027820643.1 | 1 - 2 | 72 | 114 | |
| Gene Symbol: MAP3K20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027964845.1 | XP_027820646.1 | 18 - 19 | 1708 | 114 | |
More Information
Set Membership: |
|

LOC114113152
Genomic Map