Assay Details
| Gene Symbol: LOC101119379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005510.4 | XP_004005559.3 | 5 - 6 | 583 | 81 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101119379| Gene Symbol: LOC101119379 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005510.4 | XP_004005559.3 | 5 - 6 | 583 | 81 | |