Assay Details
| Gene Symbol: CDIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010430.4 | XP_004010479.2 | 3 - 4 | 413 | 72 | |
| XM_012181160.3 | XP_012036550.2 | 3 - 4 | 413 | 72 | ||
| XM_012181161.3 | XP_012036551.2 | 3 - 4 | 413 | 72 | ||
| XM_012181162.3 | XP_012036552.2 | 3 - 4 | 413 | 72 | ||
| XM_027971655.1 | XP_027827456.1 | 3 - 4 | 413 | 72 | ||
| XM_027971656.1 | XP_027827457.1 | 3 - 4 | 127 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027971656.1 transcripts.
Set Membership: |
|

CDIN1
Genomic Map