Assay Details
| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 25 - 26 | 4134 | 92 | |
| XM_012182449.3 | XP_012037839.2 | 25 - 26 | 4137 | 92 | ||
| XM_012182451.3 | XP_012037841.2 | 25 - 26 | 4138 | 92 | ||
| Gene Symbol: LOC114116228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972738.1 | XP_027828539.1 | 3 - 4 | 427 | 92 | |

LOC101114198
Genomic Map