Assay Details
| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 27 - 28 | 4357 | 54 | |
| XM_012182449.3 | XP_012037839.2 | 27 - 28 | 4360 | 54 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114198| Gene Symbol: LOC101114198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011124.4 | XP_004011173.2 | 27 - 28 | 4357 | 54 | |
| XM_012182449.3 | XP_012037839.2 | 27 - 28 | 4360 | 54 | ||