Assay Details
| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016507.4 | XP_004016556.2 | 7 - 8 | 1088 | 77 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBLIF| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016507.4 | XP_004016556.2 | 7 - 8 | 1088 | 77 | |