Assay Details
| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016507.4 | XP_004016556.2 | 8 - 9 | 1212 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CBLIF| Gene Symbol: CBLIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016507.4 | XP_004016556.2 | 8 - 9 | 1212 | 58 | |
Set Membership: |
|