Assay Details
| Gene Symbol: GUCY1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097592.3 | XP_011952982.1 | 7 - 8 | 2322 | 101 | |
| XM_027956340.1 | XP_027812141.1 | 6 - 7 | 3480 | 101 | ||
| XM_027956341.1 | XP_027812142.1 | 7 - 8 | 2233 | 101 | ||
| XM_027956342.1 | XP_027812143.1 | 7 - 8 | 2461 | 101 | ||
More Information
Set Membership: |
|

GUCY1A1
Genomic Map