Assay Details
| Gene Symbol: MPV17L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020788.4 | XP_004020837.1 | 3 - 4 | 475 | 79 | |
| XM_027961630.1 | XP_027817431.1 | 3 - 4 | 417 | 79 | ||
More Information
Set Membership: |
|

MPV17L
Genomic Map