Assay Details
| Gene Symbol: DYDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021521.4 | XP_004021570.2 | 4 - 5 | 554 | 101 | |
| XM_027962252.1 | XP_027818053.1 | 4 - 5 | 554 | 101 | ||
| XM_027962253.1 | XP_027818054.1 | 4 - 5 | 553 | 101 | ||
More Information
Set Membership: |
|

DYDC2
Genomic Map