Assay Details
| Gene Symbol: LOC101108147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023170.4 | XP_004023219.4 | 1 - 2 | 637 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101108147| Gene Symbol: LOC101108147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023170.4 | XP_004023219.4 | 1 - 2 | 637 | 79 | |