Assay Details
| Gene Symbol: MACO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093680.2 | XP_014949166.2 | 8 - 9 | 1669 | 58 | |
| XM_027965497.1 | XP_027821298.1 | 8 - 9 | 1680 | 58 | ||
More Information
Set Membership: |
|

MACO1
Genomic Map