Assay Details
| Gene Symbol: GLTPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098599.2 | XP_014954085.2 | 1 - 2 | 279 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLTPD2| Gene Symbol: GLTPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098599.2 | XP_014954085.2 | 1 - 2 | 279 | 78 | |
Set Membership: |
|